siRNA Details
| Gene Symbol: Rbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191066.1 | NP_001177995.1 | 2 | 354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbl1| Gene Symbol: Rbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191066.1 | NP_001177995.1 | 2 | 354 | ||