siRNA Details
| Gene Symbol: Hmgn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276434.1 | NP_001263363.1 | 6 | 1254 | ||
| Gene Symbol: Hmgn5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134706.1 | NP_001128178.1 | 6 | 1335 | ||
| GenBank mRNA | BC168165.1 | - | 6 | 1237 | 8370295 | |

Hmgn5
Genomic Map