siRNA Details
| Gene Symbol: Tas2r139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080905.1 | NP_001074374.1 | 1 | 156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r139| Gene Symbol: Tas2r139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080905.1 | NP_001074374.1 | 1 | 156 | ||