siRNA Details
| Gene Symbol: Clec12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134716.1 | NP_001128188.1 | 582 | |||
| GenBank mRNA | BC168186.1 | - | 582 | 7100019 | ||
| GU357484.1 | - | 381 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec12a| Gene Symbol: Clec12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134716.1 | NP_001128188.1 | 582 | |||
| GenBank mRNA | BC168186.1 | - | 582 | 7100019 | ||
| GU357484.1 | - | 381 | ||||