siRNA Details
| Gene Symbol: Olr1500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000942.1 | NP_001000942.1 | 1 | 766 | ||
| GenBank mRNA | M64391.1 | - | 1 | 766 | ||
| Gene Symbol: Olr1873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056842.4 | XP_001056842.3 | 1 | 766 | ||

Olr1500
Genomic Map