siRNA Details
| Gene Symbol: Prss47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253500.2 | XP_006253562.1 | 3 | 891 | ||
| XM_008771453.1 | XP_008769675.1 | 3 | 892 | |||
| XM_008771454.1 | XP_008769676.1 | 3 | 614 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss47| Gene Symbol: Prss47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253500.2 | XP_006253562.1 | 3 | 891 | ||
| XM_008771453.1 | XP_008769675.1 | 3 | 892 | |||
| XM_008771454.1 | XP_008769676.1 | 3 | 614 | |||