siRNA Details
| Gene Symbol: Aup1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079899.1 | NP_001073368.1 | 11 | 1225 | ||
| GenBank mRNA | BC127544.1 | - | 11 | 1225 | 7319831 | |
| FQ228919.1 | - | 11 | 1259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aup1| Gene Symbol: Aup1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079899.1 | NP_001073368.1 | 11 | 1225 | ||
| GenBank mRNA | BC127544.1 | - | 11 | 1225 | 7319831 | |
| FQ228919.1 | - | 11 | 1259 | |||