siRNA Details
| Gene Symbol: Haus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109412.1 | NP_001102882.1 | 3 | 1524 | ||
| XM_008770361.1 | XP_008768583.1 | 2 | 2258 | |||
| XM_008770362.1 | XP_008768584.1 | 2 | 1830 | |||
| GenBank mRNA | BC162020.1 | - | 3 | 1496 | 9098415 | |

Haus3
Genomic Map