siRNA Details
| Gene Symbol: Lrrc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109413.2 | NP_001102883.1 | 3 | 646 | ||
| XM_006238402.2 | XP_006238464.1 | 4 | 1008 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc19| Gene Symbol: Lrrc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109413.2 | NP_001102883.1 | 3 | 646 | ||
| XM_006238402.2 | XP_006238464.1 | 4 | 1008 | |||