siRNA Details
| Gene Symbol: LOC680520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057542.4 | XP_001057542.2 | 2 | 1144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC680520| Gene Symbol: LOC680520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057542.4 | XP_001057542.2 | 2 | 1144 | ||