siRNA Details
| Gene Symbol: Hist1h1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109417.1 | NP_001102887.1 | 1 | 484 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h1b| Gene Symbol: Hist1h1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109417.1 | NP_001102887.1 | 1 | 484 | ||