siRNA Details
| Gene Symbol: C14orf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127393.1 | NP_001120865.1 | 3, 4 | 488 | ||
| NM_004894.2 | NP_004885.1 | 2, 3 | 219 | |||
| GenBank mRNA | AF054175.1 | - | 2, 3 | 182 | ||
| AF116639.1 | - | 215 | ||||
| AK309499.1 | - | 2, 3 | 182 | |||
| AK312099.1 | - | 2, 3 | 180 | |||
| BC000429.2 | - | 2, 3 | 171 | 2819801 | ||
| BC001944.1 | - | 2, 3 | 171 | 2819801 | ||
| BX161388.1 | - | 3, 4 | 423 | |||
| CB127906.1 | - | 3, 4 | 447 | |||

C14orf2
Genomic Map