siRNA Details
| Gene Symbol: Tet3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057850.5 | XP_001057850.2 | 5 | 4168 | ||
| XM_006236792.2 | XP_006236854.1 | 5 | 3842 | |||
| XM_006236793.2 | XP_006236855.1 | 4 | 3785 | |||
| XM_006236794.2 | XP_006236856.1 | 4 | 3211 | |||
| XM_008763094.1 | XP_008761316.1 | 6 | 3466 | |||
| XM_008763095.1 | XP_008761317.1 | 6 | 6213 | |||

Tet3
Genomic Map