siRNA Details
| Gene Symbol: Tet3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057850.5 | XP_001057850.2 | 11 | 5324 | ||
| XM_006236792.2 | XP_006236854.1 | 11 | 4998 | |||
| XM_006236793.2 | XP_006236855.1 | 10 | 4941 | |||
| XM_006236794.2 | XP_006236856.1 | 10 | 4367 | |||
| XM_008763094.1 | XP_008761316.1 | 12 | 4622 | |||
| XM_008763095.1 | XP_008761317.1 | 12 | 7369 | |||

Tet3
Genomic Map