siRNA Details
| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057911.4 | XP_001057911.2 | 18 | 1943 | ||
| XM_006257439.2 | XP_006257501.1 | 17 | 1892 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrch2| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057911.4 | XP_001057911.2 | 18 | 1943 | ||
| XM_006257439.2 | XP_006257501.1 | 17 | 1892 | |||