siRNA Details
| Gene Symbol: Ppp1r16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191072.1 | NP_001178001.1 | 6 | 817 | ||
| XM_008762383.1 | XP_008760605.1 | 6 | 901 | |||
| XM_008762384.1 | XP_008760606.1 | 6 | 1692 | |||
| XM_008762385.1 | XP_008760607.1 | 6 | 2080 | |||

Ppp1r16b
Genomic Map