siRNA Details
Gene Symbol: Ppp1r16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191072.1 | NP_001178001.1 | 10 | 1252 | ||
XM_008762383.1 | XP_008760605.1 | 10 | 1336 | |||
XM_008762384.1 | XP_008760606.1 | 10 | 2127 | |||
XM_008762385.1 | XP_008760607.1 | 10 | 2515 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
