siRNA Details
| Gene Symbol: Il1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244746.2 | XP_006244808.1 | 1 | 1631 | ||
| XM_006244750.2 | XP_006244812.1 | 1 | 1631 | |||
| XM_006244754.2 | XP_006244816.1 | 1 | 1631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1r1| Gene Symbol: Il1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244746.2 | XP_006244808.1 | 1 | 1631 | ||
| XM_006244750.2 | XP_006244812.1 | 1 | 1631 | |||
| XM_006244754.2 | XP_006244816.1 | 1 | 1631 | |||