siRNA Details
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056825.4 | XP_001056825.3 | 3 | 419 | ||
| XM_006253546.2 | XP_006253608.1 | 3 | 699 | |||
| XM_006253547.2 | XP_006253609.1 | 3 | 700 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golm1| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056825.4 | XP_001056825.3 | 3 | 419 | ||
| XM_006253546.2 | XP_006253608.1 | 3 | 699 | |||
| XM_006253547.2 | XP_006253609.1 | 3 | 700 | |||