siRNA Details
| Gene Symbol: Rpl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031065.1 | NP_112327.1 | 6 | 553 | ||
| GenBank mRNA | BC058468.1 | - | 6 | 560 | 6919792 | |
| FQ221330.1 | - | 6 | 553 | |||
| FQ221725.1 | - | 6 | 556 | |||
| FQ222317.1 | - | 6 | 558 | |||
| X93352.1 | - | 6 | 553 | |||
| Gene Symbol: LOC680700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001058455.4 | XP_001058455.1 | 2 | 562 | ||

Rpl10a
Genomic Map