siRNA Details
Gene Symbol: Nxpe4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001056892.5 | XP_001056892.3 | 4 | 1193 | ||
XM_006249027.2 | XP_006249089.1 | 5 | 1047 | |||
XM_006249028.2 | XP_006249090.1 | 4 | 1279 |
Gene Symbol: Nxpe5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769074.1 | XP_008767296.1 | 3 | 1235 |
