siRNA Details
| Gene Symbol: Nxpe5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769074.1 | XP_008767296.1 | 4 | 1870 | ||
| Gene Symbol: Nxpe4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056892.5 | XP_001056892.3 | 5 | 1828 | ||
| XM_006249027.2 | XP_006249089.1 | 6 | 1682 | |||
| XM_006249028.2 | XP_006249090.1 | 5 | 1914 | |||
| Gene Symbol: RGD1562319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769094.1 | XP_008767316.1 | 6 | 1735 | ||

Nxpe5
Genomic Map