siRNA Details
| Gene Symbol: Tmem88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109426.1 | NP_001102896.1 | 2 | 1604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem88b| Gene Symbol: Tmem88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109426.1 | NP_001102896.1 | 2 | 1604 | ||