siRNA Details
Gene Symbol: Snrpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126091.1 | NP_001119563.1 | 3, 4 | 364 | ||
GenBank mRNA | FQ214256.1 | - | 3, 4 | 304 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
