siRNA Details
| Gene Symbol: Snrpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126091.1 | NP_001119563.1 | 4 | 565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snrpf| Gene Symbol: Snrpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126091.1 | NP_001119563.1 | 4 | 565 | ||