siRNA Details
| Gene Symbol: Ccdc74a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001058720.4 | XP_001058720.2 | 4 | 2441 | ||
| XM_008768921.1 | XP_008767143.1 | 4 | 2441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc74a| Gene Symbol: Ccdc74a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001058720.4 | XP_001058720.2 | 4 | 2441 | ||
| XM_008768921.1 | XP_008767143.1 | 4 | 2441 | |||