siRNA Details
| Gene Symbol: Zcchc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257418.2 | XP_006257480.1 | 2 | 2919 | ||
| XM_008773478.1 | XP_008771700.1 | 5 | 2290 | |||
| XM_008773479.1 | XP_008771701.1 | 3 | 3122 | |||
| XM_008773480.1 | XP_008771702.1 | 2 | 620 | |||

Zcchc16
Genomic Map