siRNA Details
| Gene Symbol: Spcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191073.1 | NP_001178002.1 | 5 | 493 | ||
| GenBank mRNA | AY724480.1 | - | 612 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spcs3| Gene Symbol: Spcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191073.1 | NP_001178002.1 | 5 | 493 | ||
| GenBank mRNA | AY724480.1 | - | 612 | |||