siRNA Details
Gene Symbol: Tex13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773472.1 | XP_008771694.1 | 3 | 687 | ||
XM_008773473.1 | XP_008771695.1 | 2 | 779 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tex13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773472.1 | XP_008771694.1 | 3 | 687 | ||
XM_008773473.1 | XP_008771695.1 | 2 | 779 |