siRNA Details
| Gene Symbol: Tex13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773472.1 | XP_008771694.1 | 3 | 744 | ||
| XM_008773473.1 | XP_008771695.1 | 2 | 836 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex13b| Gene Symbol: Tex13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773472.1 | XP_008771694.1 | 3 | 744 | ||
| XM_008773473.1 | XP_008771695.1 | 2 | 836 | |||