siRNA Details
| Gene Symbol: Sf3b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126092.2 | NP_001119564.1 | 1 | 340 | ||
| GenBank mRNA | CB725243.1 | - | 1 | 340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3b5| Gene Symbol: Sf3b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126092.2 | NP_001119564.1 | 1 | 340 | ||
| GenBank mRNA | CB725243.1 | - | 1 | 340 | ||