siRNA Details
| Gene Symbol: Paxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057305.5 | XP_001057305.1 | 6 | 1260 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Paxbp1| Gene Symbol: Paxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057305.5 | XP_001057305.1 | 6 | 1260 | ||