siRNA Details
Gene Symbol: Paqr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191077.1 | NP_001178006.1 | 5, 6 | 571 | ||
XM_006232755.2 | XP_006232817.1 | 5, 6 | 1573 | |||
XM_008761210.1 | XP_008759432.1 | 4, 5 | 1648 | |||
XM_008761211.1 | XP_008759433.1 | 4, 5 | 1516 | |||
XM_008761212.1 | XP_008759434.1 | 4, 5 | 753 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
