siRNA Details
| Gene Symbol: LOC681066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060160.5 | XP_001060160.4 | 1 | 790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC681066| Gene Symbol: LOC681066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060160.5 | XP_001060160.4 | 1 | 790 | ||