siRNA Details
| Gene Symbol: AVPR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000707.3 | NP_000698.1 | 1 | 846 | ||
| GenBank mRNA | AF030512.1 | - | 1 | 811 | ||
| AF101726.1 | - | 1 | 811 | |||
| D31833.1 | - | 1 | 895 | |||
| EU432111.1 | - | 1 | 688 | |||
| L37112.1 | - | 1 | 782 | |||
More Information
Set Membership: |
|

AVPR1B
Genomic Map