siRNA Details
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246025.2 | XP_006246087.1 | 14 | 4645 | ||
| XM_006246026.2 | XP_006246088.1 | 14 | 4645 | |||
| XM_008767590.1 | XP_008765812.1 | 14 | 4642 | |||
| XM_008767591.1 | XP_008765813.1 | 14 | 4249 | |||
| Gene Symbol: Dnase1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057412.3 | XP_001057412.4 | 1 | 305 | ||
| XM_006246041.1 | XP_006246103.1 | 1 | 305 | |||
| XM_006246042.1 | XP_006246104.1 | 1 | 305 | |||

E4f1
Genomic Map