siRNA Details
| Gene Symbol: Ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013053.1 | NP_037185.1 | 5, 6 | 757 | ||
| GenBank mRNA | BC062409.1 | - | 4, 5 | 727 | 5623850 | |
| D17614.1 | - | 5, 6 | 757 | |||
| FQ219995.1 | - | 1, 2 | 44 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ywhaq| Gene Symbol: Ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013053.1 | NP_037185.1 | 5, 6 | 757 | ||
| GenBank mRNA | BC062409.1 | - | 4, 5 | 727 | 5623850 | |
| D17614.1 | - | 5, 6 | 757 | |||
| FQ219995.1 | - | 1, 2 | 44 | |||