siRNA Details
Gene Symbol: MGC94891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044277.1 | NP_001037742.2 | 9 | 1399 | ||
GenBank mRNA | BC083819.1 | - | 9 | 1399 | 7111103 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MGC94891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044277.1 | NP_001037742.2 | 9 | 1399 | ||
GenBank mRNA | BC083819.1 | - | 9 | 1399 | 7111103 |