siRNA Details
| Gene Symbol: Dmtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053693.1 | NP_446145.1 | 15 | 1982 | ||
| XM_006235993.1 | XP_006236055.1 | 15 | 2044 | |||
| GenBank mRNA | AF352171.1 | - | 1 | 118 | ||
| BC081880.1 | - | 15 | 1982 | 7108159 | ||
| Gene Symbol: LOC681224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060830.3 | XP_001060830.1 | 1 | 863 | ||

Dmtf1
Genomic Map