siRNA Details
| Gene Symbol: T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161835.1 | NP_001155307.1 | 11 | 1248 | ||
| XM_006227933.2 | XP_006227995.1 | 11 | 2210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

T2| Gene Symbol: T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161835.1 | NP_001155307.1 | 11 | 1248 | ||
| XM_006227933.2 | XP_006227995.1 | 11 | 2210 | |||