siRNA Details
| Gene Symbol: LOC681300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257539.2 | XP_006257601.1 | 1 | 923 | ||
| Gene Symbol: Cxxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007628.2 | NP_001007629.1 | 1 | 1007 | ||
| GenBank mRNA | BC083622.1 | - | 1 | 1007 | 7134100 | |

LOC681300
Genomic Map