siRNA Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122781.3 | NP_001116253.3 | 2 | 232 | ||
| XM_006242354.2 | XP_006242416.1 | 2 | 295 | |||
| GenBank mRNA | BC159426.1 | - | 2 | 213 | 9034024 | |
| CB778986.1 | - | 2 | 254 | |||

Letmd1
Genomic Map