siRNA Details
| Gene Symbol: LOC681382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001061496.2 | XP_001061496.1 | 1 | 156 | ||
| Gene Symbol: Cenpw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001246319.1 | NP_001233248.1 | 2 | 257 | ||
| GenBank mRNA | BC129114.1 | - | 2 | 257 | 7112235 | |

LOC681382
Genomic Map