siRNA Details
| Gene Symbol: LOC681383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109441.1 | NP_001102911.1 | 5 | 595 | ||
| XM_006251618.2 | XP_006251680.1 | 5 | 604 | |||
| XM_008770472.1 | XP_008768694.1 | 4 | 908 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC681383| Gene Symbol: LOC681383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109441.1 | NP_001102911.1 | 5 | 595 | ||
| XM_006251618.2 | XP_006251680.1 | 5 | 604 | |||
| XM_008770472.1 | XP_008768694.1 | 4 | 908 | |||