siRNA Details
| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003690.1 | NP_001003690.1 | 3 | 591 | ||
| NM_014628.2 | NP_055443.1 | 2 | 323 | |||
| GenBank mRNA | AK297172.1 | - | 2 | 344 | ||
| AL537692.3 | - | 3 | 593 | |||
| BC002904.2 | - | 2 | 293 | 3945348 | ||
| BE297042.1 | - | 3 | 519 | |||
| BI255998.1 | - | 2 | 310 | |||
| BU845738.1 | - | 3 | 628 | |||
| CB991410.1 | - | 2 | 350 | |||
| CR456893.1 | - | 2 | 287 | |||
| D14811.1 | - | 2 | 290 | |||

MAD2L1BP
Genomic Map