siRNA Details
| Gene Symbol: Nsa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134415.2 | NP_599242.2 | 2 | 272 | ||
| GenBank mRNA | AF158379.1 | - | 2 | 280 | ||
| BC098657.1 | - | 2 | 263 | 7378282 | ||
| DQ354588.1 | - | 2 | 262 | |||
| FQ212851.1 | - | 2 | 284 | |||
| FQ214845.1 | - | 2 | 291 | |||
| FQ225571.1 | - | 2 | 285 | |||
| FQ226635.1 | - | 2 | 299 | |||
| FQ229542.1 | - | 2 | 290 | |||
| Y17325.1 | - | 2 | 249 | |||

Nsa2
Genomic Map