siRNA Details
| Gene Symbol: Rnf133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044278.1 | NP_001037743.1 | 1 | 778 | ||
| XM_006236144.2 | XP_006236206.1 | 2 | 2690 | |||
| GenBank mRNA | BC079249.1 | - | 1 | 778 | 7133029 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf133| Gene Symbol: Rnf133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044278.1 | NP_001037743.1 | 1 | 778 | ||
| XM_006236144.2 | XP_006236206.1 | 2 | 2690 | |||
| GenBank mRNA | BC079249.1 | - | 1 | 778 | 7133029 | |