siRNA Details
| Gene Symbol: Prr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173428.1 | NP_001166899.1 | 2 | 473 | ||
| XM_006228478.2 | XP_006228540.1 | 2 | 468 | |||
| XM_006228479.2 | XP_006228541.1 | 2 | 380 | |||
| XM_006228480.2 | XP_006228542.1 | 2 | 468 | |||
| XM_008758985.1 | XP_008757207.1 | 2 | 576 | |||
| XM_008758986.1 | XP_008757208.1 | 2 | 578 | |||
| XM_008758987.1 | XP_008757209.1 | 2 | 633 | |||
| GenBank mRNA | BC168703.1 | - | 2 | 472 | 7455407 | |

Prr19
Genomic Map