siRNA Details
| Gene Symbol: Ndufb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109443.1 | NP_001102913.1 | 1, 2 | 242 | ||
| GenBank mRNA | FQ218220.1 | - | 1, 2 | 187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb10| Gene Symbol: Ndufb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109443.1 | NP_001102913.1 | 1, 2 | 242 | ||
| GenBank mRNA | FQ218220.1 | - | 1, 2 | 187 | ||