siRNA Details
Gene Symbol: H2afv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106019.1 | NP_001099489.1 | 3 | 265 | ||
XM_006251387.2 | XP_006251449.1 | 3 | 603 |
Gene Symbol: LOC685069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237209.2 | XP_006237271.1 | 1 | 191 |
Gene Symbol: LOC689408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001070681.5 | XP_001070681.4 | 1 | 196 |
Gene Symbol: RGD1560813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751821.2 | XP_003751869.1 | 1 | 219 |
